Order Entry
United Kingdom
ContactUsLinkComponent
Precision therm., low temp., similar DIN, -50+50:0,1C, solid stem, white backed, toluene filled, 580x6-7mm, with ring at top, suit able for government calibration 1 * 1 items
Catalog # AMARL33108-TOL
Supplier:  Amarell
Precision therm., low temp., similar DIN, -50+50:0,1C, solid stem, white backed, toluene filled, 580x6-7mm, with ring at top, suit able for government calibration 1 * 1 items
Catalog # AMARL33108-TOL
Supplier:  Amarell
Frequently Bought Together