Order Entry
United Kingdom
ContactUsLinkComponent
Precision therm., low temp., similar DIN, -50+50:0,5C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suit able for government verification 1 * 1 items
  AMARL33098-TOL
 :  Amarell
Precision therm., low temp., similar DIN, -50+50:0,5C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suit able for government verification 1 * 1 items
  AMARL33098-TOL
 :  Amarell
Frequently Bought Together