Precision therm., low temp., DIN 12778, -100+30:1C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suitabl e for government verification 1 * 1 items
Catalog # AMARL33092-TOL
Supplier: Amarell
Precision therm., low temp., DIN 12778, -100+30:1C, solid stem, white backed, toluene filled, 300x6-7mm, with ring at top, suitabl e for government verification 1 * 1 items
Catalog # AMARL33092-TOL
Supplier: Amarell
Frequently Bought Together
VWR®, Digital Timer, Two Channels
Supplier: VWR Collection
Beakers, low form
Supplier: KARTELL
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Accessories for water purification systems, arium®
Supplier: Sartorius
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
DURAN® Screw Caps with Aperture
Supplier: DWK Life Sciences
Digital timer
Supplier: GLASWARENFABRIK KARL HECHT
Accessories for marco
Supplier: Brand
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for VWR® Pipette Controllers
Supplier: VWR Collection
VWR®, Digital Timer, Three Channels, with Clock
Supplier: VWR Collection
Cartridges for Milli-Q®
Supplier: Merck
Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF