Precision therm., high temp., similiar DIN, +200+480:1°C, enclosed scale, cap. colourless, total immersion, gallium filled, 450mm lo ng, diameter 9-10mm, durable pigment, Richter-top finish, suitable for government verification 1 * 1 items
Catalog # AMARL29720-GA
Supplier: Amarell
Frequently Bought Together