Order Entry
United Kingdom
ContactUsLinkComponent
Precision thermometer, similar DIN, 0+50:0,1C, solid stem, white backed, red special liquid filled, 420x6-7mm, durable pigment, sui table for government calibration 1 * 1 items
Catalog # AMARL27374-FL
Supplier:  Amarell
Precision thermometer, similar DIN, 0+50:0,1C, solid stem, white backed, red special liquid filled, 420x6-7mm, durable pigment, sui table for government calibration 1 * 1 items
Catalog # AMARL27374-FL
Supplier:  Amarell
Frequently Bought Together