Order Entry
United Kingdom
ContactUsLinkComponent
Precision thermometer, similar DIN, -5+200:0,2°C, solid stem, white backed, red special liquid, 550x6-7mm, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARL27362-FL
Supplier:  Amarell
Precision thermometer, similar DIN, -5+200:0,2°C, solid stem, white backed, red special liquid, 550x6-7mm, durable pigment, suitable for government calibration 1 * 1 items
Catalog # AMARL27362-FL
Supplier:  Amarell
Frequently Bought Together