Order Entry
United Kingdom
ContactUsLinkComponent
Precision thermometer, similar DIN, -5+100:0,2C, solid stem, white backed, capillary tube specially coated inside, with non-wettin g blue special liquid, 420x6-7mm, durable pigment, null
Catalog # AMARL27358-CB
Supplier:  Amarell
Precision thermometer, similar DIN, -5+100:0,2C, solid stem, white backed, capillary tube specially coated inside, with non-wettin g blue special liquid, 420x6-7mm, durable pigment, null
Catalog # AMARL27358-CB
Supplier:  Amarell
Frequently Bought Together