Thermometer similar to AFNOR, STL/0,1/-5/25, -5+25:0,1°C, white backed, capillary tube specially coated inside, with non-wetting bl ue special liquid, 375mm long, total immersion, durable pigment, null
Catalog # AMARA411002-CB
Supplier: Amarell
Frequently Bought Together