Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Rat EGF

2 µG
Catalog # APOSBITP2077-2UG
Supplier:   Apollo Scientific
Rat EGF
Catalog # APOSBITP2077-2UG
Supplier:  Apollo Scientific
CAS Number:  
Rat EGF

2 µG

CAS Number:  
Price
£214.00
Log in for availability
1 * 2 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    2 µG
  • Cat. No.:
    APOSBITP2077-2UG
  • Protein/peptide name:
    EGF
  • Protein synonyms:
    Epidermal Growth Factor
  • Species:
    Rat

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.