3-Phenoxypropionic acid
Bottle, 5 g
Catalog # APOSOR13898-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 3-Penoxypropionic acid, 3-Phenoxypropanoic acid
- CAS Number:7170-38-9
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002769
- Molecular Weight:166.18 g/mol
- Boiling Pt:235…245 °C
- Melting Pt:98…100 °C
- Storage Temperature:Ambient
- EINECS:230-518-4
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



