4'-Hydroxyacetophenone
Bottle, 500 g
Catalog # APOSOR11052-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:99-93-4
- Formula:C₈H₈O₂
- MDL Number:MFCD00002359
- Molecular Weight:136.15 g/mol
- Boiling Pt:147…148 °C/3mmHg
- Melting Pt:108…110 °C
- Density:1.109 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:202-802-8
Specifications
Specifications
- Pack type:Bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



