4-Hydroxy-8-methoxyquinoline
Bottle, 5 g
Catalog # APOSOR307979-5G
Supplier: Apollo Scientific
4-Hydroxy-8-methoxyquinoline
Catalog # APOSOR307979-5G
Supplier: Apollo Scientific
CAS Number: 21269-34-1
Jump to:
Product Details & Documents
- CAS Number:21269-34-1
- Formula:C₁₀H₉NO₂
- MDL Number:MFCD02684163
- Molecular Weight:175.184 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



