1-Iodo-2-naphthol
Bottle, 5 g
Catalog # APOSOR30346-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Hydroxy-1-iodonaphthalene
- CAS Number:2033-42-3
- Formula:C₁₀H₇IO
- MDL Number:MFCD00046371
- Molecular Weight:270.07 g/mol
- Boiling Pt:307 °C/760 mmHg
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



