Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
N-Hydroxysuccinimide 99%

N-Hydroxysuccinimide 99%

Bottle, 1 kg
Catalog # APOSOR28434-1KG
Irritant
Supplier:   Apollo Scientific
N-Hydroxysuccinimide 99%
Catalog # APOSOR28434-1KG
Supplier:  Apollo Scientific
CAS Number:  6066-82-6
N-Hydroxysuccinimide 99%

Bottle, 1 kg

CAS Number:  6066-82-6
Price
£72.60
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu
  • CAS Number:6066-82-6
  • Formula:C₄H₅NO₃
  • MDL Number:MFCD00005516
  • Molecular Weight:115.09 g/mol
  • Melting Pt:94…99 °C
  • Storage Temperature:Ambient
  • EINECS:228-001-3

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications