Perfluoro(methylcyclohexane)
Bottle, 250 g
Catalog # APOSPC4381N-250G
Supplier: Apollo Scientific
Perfluoro(methylcyclohexane)
Catalog # APOSPC4381N-250G
Supplier: Apollo Scientific
CAS Number: 355-02-2
Jump to:
Product Details & Documents
Synonyms: FlutecR PP2
- CAS Number:355-02-2
- Formula:C₇F₁₄
- MDL Number:MFCD00001468
- Molecular Weight:350.05 g/mol
- Boiling Pt:76 °C
- Density:1.788 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:206-573-5
Specifications
Specifications
- Pack type:Bottle
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



