Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

3-Fluoropyrocatechol 99%

Bottle, 25 g
Catalog # APOSPC3728D-25G
Corrosive
Irritant
Supplier:   Apollo Scientific
3-Fluoropyrocatechol 99%
Catalog # APOSPC3728D-25G
Supplier:  Apollo Scientific
CAS Number:  363-52-0
3-Fluoropyrocatechol 99%

Bottle, 25 g

CAS Number:  363-52-0
Price
£101.00
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Fluorocatechol, 3-Fluorobenzene-1,2-diol, 3-Fluoro-1,2-dihydroxybenzene
  • CAS Number:363-52-0
  • Formula:C₆H₅FO₂
  • MDL Number:MFCD00042582
  • Molecular Weight:128.1 g/mol
  • Melting Pt:71…73 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications