Lead ACS
Bottle, 2 kg
Catalog # APOSIN2182-2KG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:7439-92-1
- Formula:Pb
- MDL Number:MFCD00134050
- Molecular Weight:207.2 g/mol
- Boiling Pt:1740 °C
- Melting Pt:327.43 °C
- Density:11.34 g/cm³ (25 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:2 kg



