Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Iodophenol 98%

2-Iodophenol 98%

Bottle, 25 g
Catalog # APOSOR4682-25G
Irritant
Supplier:   Apollo Scientific
2-Iodophenol 98%
Catalog # APOSOR4682-25G
Supplier:  Apollo Scientific
CAS Number:  533-58-4
2-Iodophenol 98%

Bottle, 25 g

CAS Number:  533-58-4
Price
£19.80
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:533-58-4
  • Formula:C₆H₅IO
  • MDL Number:MFCD00013963
  • Molecular Weight:220.01 g/mol
  • Boiling Pt:186…187 °C/160mmHg
  • Melting Pt:37…43 °C
  • Density:1.947 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:208-569-9
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.