G418 disulphate
Bottle, 5 g
Catalog # APOSBIG0175-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Aminoglycoside antibiotic.
Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
- CAS Number:108321-42-2
- Formula:C₂₀H₄₄N₄O₁₈S₂
- MDL Number:MFCD00058314
- Molecular Weight:692.72 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Pack type:Bottle
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



