Order Entry
United Kingdom
ContactUsLinkComponent
True colour LED light source for BX53F2 microscope frames. LED features unique colour rendering index (>96) in transmission observations similar like halogen light source. Intensity higher than 100W halogen light. Expected lifetime of 50,000 h. Power 1 * 1 items
Catalog # OLYMN5685300
Supplier:  OLYMPUS OPTICAL
True colour LED light source for BX53F2 microscope frames. LED features unique colour rendering index (>96) in transmission observations similar like halogen light source. Intensity higher than 100W halogen light. Expected lifetime of 50,000 h. Power 1 * 1 items
Catalog # OLYMN5685300
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together