Order Entry
United Kingdom
ContactUsLinkComponent
Facilitates gases other than hydrogen to be used on the H-Cube Pro supplied through either a gas cylinder or a gas generator. The gases that can be utilized through the Gas Module are compressed air/oxygen/carbon monoxide/ethylene/SynGas/methane/etha 1 * 1 items
Catalog # IBSATHS09027
Supplier:  IBS Stock Items
Facilitates gases other than hydrogen to be used on the H-Cube Pro supplied through either a gas cylinder or a gas generator. The gases that can be utilized through the Gas Module are compressed air/oxygen/carbon monoxide/ethylene/SynGas/methane/etha 1 * 1 items
Catalog # IBSATHS09027
Supplier:  IBS Stock Items
Frequently Bought Together