Order Entry
United Kingdom
ContactUsLinkComponent
SPECTRAPLATE-96 HB, CLEAR 96-WELL MICROPLATE WITH HIGH PROTEIN BINDING CAPACITY 1 * 50 items
Catalog # PERK6005600
Supplier:  PerkinElmer
SPECTRAPLATE-96 HB, CLEAR 96-WELL MICROPLATE WITH HIGH PROTEIN BINDING CAPACITY 1 * 50 items
Catalog # PERK6005600
Supplier:  PerkinElmer
Frequently Bought Together