4-Nitrophenyl phosphate 97%
Glass bottle, 10 g
Catalog # ACRO455010100
Supplier: Thermo Fisher Scientific
4-Nitrophenyl phosphate 97%
Catalog # ACRO455010100
Supplier: Thermo Fisher Scientific
CAS Number: 330-13-2
Jump to:
Product Details & Documents
Synonyms: 4-Nitrophenyl dihydrogen phosphate, pNpp, p-Nitrophenyl phosphate
- CAS Number:330-13-2
- Formula:C₆H₆NO₆P
- Molecular Weight:219.09 g/mol
- Boiling Pt:458 °C
- Melting Pt:153 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



