Order Entry
United Kingdom
ContactUsLinkComponent
Clear round bottom, polystyrene, 384-well, individually packed with lid, sterile, for 3D spheroid formation and measurement 1 * 20 items
Catalog # NEXCULA-96U-020
Clear round bottom, polystyrene, 384-well, individually packed with lid, sterile, for 3D spheroid formation and measurement 1 * 20 items
Catalog # NEXCULA-96U-020
Frequently Bought Together