Cell viability kit contains three reagents: Calcein AM, PI, and Hoechst. Live cells are labled with green fluorescing calcein, dead cells are labeled with red fluorescing PI, and total nucleated cell number is determined using Hoechst. 1 * 1 items
Catalog # NEXCCSK-V0001-1
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together