Calcein AM is a cell permeable, non-fluorescent compound. Upon crossing the cell membrane, it is rapidly hydrolyzed converting the n on-fluorescent calcein AM to a strongly green fluorescing Calcein. Cells that are unable to convert calcein AM to cal 1 * 200 µl
Catalog # NEXCCS1-0119
Supplier: NEXCELOM BIOSCIENCE
Frequently Bought Together