Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Accessories for Cellometer®
Accessories for Cellometer®
Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
Catalog # NEXCCHT4-PD300-003
Supplier:   NEXCELOM BIOSCIENCE
Accessories for Cellometer®
Catalog # NEXCCHT4-PD300-003
Accessories for Cellometer®
Accessories for Cellometer®
Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
Price
Inquire For Price
Log in for availability
1 * 500 items
Product Details & Documents
Specifications
  • Description:
    Disposable counting chambers (300 µm) for cells greater than 80 µm compatible with Auto 2000, packed in a case of 500 slides for 1000 counts
  • Seq:
    904