Order Entry
United Kingdom
ContactUsLinkComponent
Vision CBA Analysis System: Includes Vision HSL instrument, 2 fluorescence optics modules,  2 licenses of Vision CBA software (one f or instrument, 1 at remote workstation), automated controller, USB cable, power supply, box of 75 slides, 24 month wa 1 * 1 items
  NEXCVCBA-SYS1-10
Vision CBA Analysis System: Includes Vision HSL instrument, 2 fluorescence optics modules,  2 licenses of Vision CBA software (one f or instrument, 1 at remote workstation), automated controller, USB cable, power supply, box of 75 slides, 24 month wa 1 * 1 items
  NEXCVCBA-SYS1-10
Frequently Bought Together