GREY/GREY SANTOPRENE 1.30 MM ID PKG.12 1 * 12 items
Catalog # PERCN8152415
Supplier: PerkinElmer
Frequently Bought Together
Gas Washing Bottle with Filter and Thread GL 45
Supplier: ROBU GLASFILTER
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Tubing for peristaltic pumps
Supplier: PerkinElmer