Order Entry
United Kingdom
ContactUsLinkComponent
Model SVI2S15 (Body only) ¼” BSP 316 stainless steel rear connection Uniflux VA flowmeter C/w integral fine control valve 1 * 1 items
Catalog # IBSASVI2S15
Supplier:  IBS Stock Items
Model SVI2S15 (Body only) ¼” BSP 316 stainless steel rear connection Uniflux VA flowmeter C/w integral fine control valve 1 * 1 items
Catalog # IBSASVI2S15
Supplier:  IBS Stock Items
Frequently Bought Together