Order Entry
United Kingdom
ContactUsLinkComponent
Refrigerator ES Series 05REETSA Capacity (l): 155,7 Temperature (°C): +1 to +12 Manual temperature controller Defrost: manual Explosion-Proof Refrigerator D×W×H ext. (cm) 60 x 60 x 85.1 cm Weight (kg) 58 Electrical power: 115V/60Hz 1 * 1 items
Catalog # REVC05REETSA
Supplier:  REVCO TECHNOLOGIES
Refrigerator ES Series 05REETSA Capacity (l): 155,7 Temperature (°C): +1 to +12 Manual temperature controller Defrost: manual Explosion-Proof Refrigerator D×W×H ext. (cm) 60 x 60 x 85.1 cm Weight (kg) 58 Electrical power: 115V/60Hz 1 * 1 items
Catalog # REVC05REETSA
Supplier:  REVCO TECHNOLOGIES
Frequently Bought Together