Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 8 MG/ML (19 MM) WATER LOVASTATIN IS AN INHIBITOR OF HMG-C OA REDUCTASE WITH IC50 OF 3.4 NM, USED FOR LOWERING CHOLESTEROL (HYPOLIPIDEMIC AGENT). TARGET: HMG-COA REDUCTASELOVAS 1 * 50 mg
Catalog # MDEXHY-N0504-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 8 MG/ML (19 MM) WATER LOVASTATIN IS AN INHIBITOR OF HMG-C OA REDUCTASE WITH IC50 OF 3.4 NM, USED FOR LOWERING CHOLESTEROL (HYPOLIPIDEMIC AGENT). TARGET: HMG-COA REDUCTASELOVAS 1 * 50 mg
Catalog # MDEXHY-N0504-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together