Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 20 MG/ML (37 MM) WATER 53 MG/ML (98 MM)NAFAMOSTAT MESYLATE, A SYNTHETIC S ERINE PROTEASE INHIBITOR, IS AN ANTICOAGULANT.TARGET: SERINE PROTEASETRANILAST (FUT-175) IS AN ANTIALLERGIC DRUG FOR 1 * 50 mg
Catalog # MDEXHY-B0190A-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 20 MG/ML (37 MM) WATER 53 MG/ML (98 MM)NAFAMOSTAT MESYLATE, A SYNTHETIC S ERINE PROTEASE INHIBITOR, IS AN ANTICOAGULANT.TARGET: SERINE PROTEASETRANILAST (FUT-175) IS AN ANTIALLERGIC DRUG FOR 1 * 50 mg
Catalog # MDEXHY-B0190A-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together