Order Entry
United Kingdom
ContactUsLinkComponent
SOLUBILITY: 10 MM IN DMSOA939572 (SCD1 INHIBITOR) IS A POTENT, AND ORALLY BIOAVAILABLE PIPERIDINE-ARYL UREA-BASED SCD1 INHIBITOR WITH IC50 VALUES OF <4 NM AND 37 NM FOR MSCD1 AND HSCD1 RESPECTIVELY.IC50 VALUE: <4 NM (MSCD1) 37 NM (HSCD1) [1]TAR 1 * 2 mg
Catalog # MDEXHY-50709-2MG
Supplier:  MEDCHEMEXPRESS
SOLUBILITY: 10 MM IN DMSOA939572 (SCD1 INHIBITOR) IS A POTENT, AND ORALLY BIOAVAILABLE PIPERIDINE-ARYL UREA-BASED SCD1 INHIBITOR WITH IC50 VALUES OF <4 NM AND 37 NM FOR MSCD1 AND HSCD1 RESPECTIVELY.IC50 VALUE: <4 NM (MSCD1) 37 NM (HSCD1) [1]TAR 1 * 2 mg
Catalog # MDEXHY-50709-2MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together