Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: 15 MG/MLT-863(DGAT-1 INHIBITOR) IS AN ORALLY ACTIVE, SELECTIVE AND POTENT DGAT1 (ACYL-COA:DIACYLGLYCEROL ACYLTRANSFERASE 1) INHIBITOR THAT INTERACTS WITH THE ACYL-COA BINDING SITE OF DGAT1, I 1 * 5 mg
Catalog # MDEXHY-32219-5MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: 15 MG/MLT-863(DGAT-1 INHIBITOR) IS AN ORALLY ACTIVE, SELECTIVE AND POTENT DGAT1 (ACYL-COA:DIACYLGLYCEROL ACYLTRANSFERASE 1) INHIBITOR THAT INTERACTS WITH THE ACYL-COA BINDING SITE OF DGAT1, I 1 * 5 mg
Catalog # MDEXHY-32219-5MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together