LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥9MG/ML WATER GSK461364 IS A POTENT SMALL MOLECU LE POLO-LIKE KINASE 1 (PLK1) INHIBITOR WITH A KI OF 2.2 NM. IC50 VALUE: 2.2 NM(KI)TARGET: PLK1IN VITRO: GSK461364 IS 1 * 5 mg
Catalog # MDEXHY-50877-5MG
Supplier: MEDCHEMEXPRESS
Frequently Bought Together