Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOKJ PYR 9 IS A NOVEL SMALL-MOLECULE INHIBITOR OF MYC, KJ-PYR-9, FRO M A KR?HNKE PYRIDINE LIBRARY. THE KD OF KJ-PYR-9 FOR MYC IN VITRO IS 6.5 ± 1.0 NM, AS DETERMINED BY BACKSCATTERING IN 1 * 50 mg
Catalog # MDEXHY-19735-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOKJ PYR 9 IS A NOVEL SMALL-MOLECULE INHIBITOR OF MYC, KJ-PYR-9, FRO M A KR?HNKE PYRIDINE LIBRARY. THE KD OF KJ-PYR-9 FOR MYC IN VITRO IS 6.5 ± 1.0 NM, AS DETERMINED BY BACKSCATTERING IN 1 * 50 mg
Catalog # MDEXHY-19735-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together