Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 31 MG/MLACALABRUTINIB (ACP-196) IS A SECOND-GENERATION, SELECTIVE, IRRE VERSIBLE INHIBITOR OF BTK (BRUTON'S TYROSINE KINASE) THAT HAS IMPROVED PHARMACOLOGIC FEATURES, INCLUDING FAVORABLE PL 1 * 50 mg
Catalog # MDEXHY-17600-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 31 MG/MLACALABRUTINIB (ACP-196) IS A SECOND-GENERATION, SELECTIVE, IRRE VERSIBLE INHIBITOR OF BTK (BRUTON'S TYROSINE KINASE) THAT HAS IMPROVED PHARMACOLOGIC FEATURES, INCLUDING FAVORABLE PL 1 * 50 mg
Catalog # MDEXHY-17600-50MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together