Order Entry
United Kingdom
ContactUsLinkComponent
SOLUBILITY: SOLUBLE IN DMSOSBI-0640756(SBI-756) IS A FIRST-IN-CLASS INHIBITOR THAT TARGETS EIF4G1 AND DISRUPTS THE EIF4F COMPLEX . SBI-756 ALSO SUPPRESSED AKT AND NF-ΚB SIGNALING.1) SBI-756 INHIBITED THE GROWTH OF NRAS, BRAF, AND NF1-MUTANT MELAN 1 * 25 mg
Catalog # MDEXHY-19560-25MG
Supplier:  MEDCHEMEXPRESS
SOLUBILITY: SOLUBLE IN DMSOSBI-0640756(SBI-756) IS A FIRST-IN-CLASS INHIBITOR THAT TARGETS EIF4G1 AND DISRUPTS THE EIF4F COMPLEX . SBI-756 ALSO SUPPRESSED AKT AND NF-ΚB SIGNALING.1) SBI-756 INHIBITED THE GROWTH OF NRAS, BRAF, AND NF1-MUTANT MELAN 1 * 25 mg
Catalog # MDEXHY-19560-25MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together