Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOROCAGLAMIDE (ROCAGLAMIDE A) A NATURALLY OCCURRING PRODUCT ISOLATED FROM THE GENUS AGLAIA, WITH THE IC50 OF ~50 NM FOR THE HEAT SHOCK REPORTER.IC50 VALUE: 50 NM (FOR THE HEAT SHOCK REP 1 * 10 mg
Catalog # MDEXHY-19356-10MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOROCAGLAMIDE (ROCAGLAMIDE A) A NATURALLY OCCURRING PRODUCT ISOLATED FROM THE GENUS AGLAIA, WITH THE IC50 OF ~50 NM FOR THE HEAT SHOCK REPORTER.IC50 VALUE: 50 NM (FOR THE HEAT SHOCK REP 1 * 10 mg
Catalog # MDEXHY-19356-10MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together