Order Entry
United Kingdom
ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 51 MG/MLABT-199 (GDC-0199) IS A BCL-2-SELECTIVE INHIBITOR WITH KI OF 4800-FOLD MORE SELECTIVE VERSUS BCL-XL AND BCL-W, AND NO ACTIVITY TO MCL-1. IC50 VALUE: <0.01 NM (KI)TARGET: 1 * 100 mg
Catalog # MDEXHY-15531-100MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 51 MG/MLABT-199 (GDC-0199) IS A BCL-2-SELECTIVE INHIBITOR WITH KI OF 4800-FOLD MORE SELECTIVE VERSUS BCL-XL AND BCL-W, AND NO ACTIVITY TO MCL-1. IC50 VALUE: <0.01 NM (KI)TARGET: 1 * 100 mg
Catalog # MDEXHY-15531-100MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together