Order Entry
United Kingdom
ContactUsLinkComponent
SOLUBILITY: DMSO 95 MG/ML WATER 95 MG/ML ETHANOL RIGOSERTIB (ON-01910) IS A NON-ATP-COMPETITIVE INHIBITOR OF PLK1 WITH IC50 OF 9 NM SHOWS 30-FOLD GREATER SELECTIVITY AGAINST PLK2 AND NO ACTIVITY TO PLK3.IC50 VALUE: 9 NM [1] TARGET: PL 1 * 5 mg
Catalog # MDEXHY-12037A-5MG
Supplier:  MEDCHEMEXPRESS
SOLUBILITY: DMSO 95 MG/ML WATER 95 MG/ML ETHANOL RIGOSERTIB (ON-01910) IS A NON-ATP-COMPETITIVE INHIBITOR OF PLK1 WITH IC50 OF 9 NM SHOWS 30-FOLD GREATER SELECTIVITY AGAINST PLK2 AND NO ACTIVITY TO PLK3.IC50 VALUE: 9 NM [1] TARGET: PL 1 * 5 mg
Catalog # MDEXHY-12037A-5MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together