Order Entry
United Kingdom
ContactUsLinkComponent
SOLUBILITY: DMSO: 10 MMMK-4101 IS A POTENT SMO INHIBITOR OF THE HEDGEHOG PATHWAY, HIGHLY ACTIVE AGAINST MEDULLOBLASTOMA AND BASA L CELL CARCINOMA.TARGET:SMOIN VIVO: MK-4101 WAS HIGHLY EFFICACIOUS AGAINST PRIMARY MEDULLOBLASTOMA AND BCC DEVELOPING 1 * 5 mg
Catalog # MDEXHY-100036-5MG
Supplier:  MEDCHEMEXPRESS
SOLUBILITY: DMSO: 10 MMMK-4101 IS A POTENT SMO INHIBITOR OF THE HEDGEHOG PATHWAY, HIGHLY ACTIVE AGAINST MEDULLOBLASTOMA AND BASA L CELL CARCINOMA.TARGET:SMOIN VIVO: MK-4101 WAS HIGHLY EFFICACIOUS AGAINST PRIMARY MEDULLOBLASTOMA AND BCC DEVELOPING 1 * 5 mg
Catalog # MDEXHY-100036-5MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together