BUFFER PH 7,00 1 * 1 L
Catalog # ENDRCY20-E10A1
Supplier: Endress+Hauser
Frequently Bought Together
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Nitrite 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Potassium iodide 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1070502
Supplier: VWR Chemicals
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
NexION Setup Solution 1 ug/L 500mL 1 * 1 items
Supplier: PerkinElmer
Methyl-4-hydroxybenzoate
Supplier: Merck
Turbidity standards, Stablcal®
Supplier: Hach
Supelco® Organic Reference Standard, 1H-1,2,4-Triazole
Supplier: Merck