Order Entry
United Kingdom
ContactUsLinkComponent
Inert gas connection - with gas inlet and outlet, Ø 10 mm, incl. additional measures for the greatest possible impermeability to gas not in conjunction with access port or interior lighting option 1 * 1 items
Catalog # BIND8012-1694
Supplier:  Binder
Inert gas connection - with gas inlet and outlet, Ø 10 mm, incl. additional measures for the greatest possible impermeability to gas not in conjunction with access port or interior lighting option 1 * 1 items
Catalog # BIND8012-1694
Supplier:  Binder
Frequently Bought Together