Order Entry
United Kingdom
ContactUsLinkComponent
Switched-mode Power Supply Power Amplifier QSC PL 340, Power amp, 19 /2HE, 2x 1250W/4 Ohm, 2x 800W/8 Ohm, 2x 2000W/2 Ohm, sym. XLR Inputs,Speaker Twist and Binding-Post Outputs, Gnd-lift, Protections, fan, 11 kg, 35,6cm depth (from front mounting rai 1 * 1 items
Catalog # IBSA139139
Supplier:  IBS Stock Items
Switched-mode Power Supply Power Amplifier QSC PL 340, Power amp, 19 /2HE, 2x 1250W/4 Ohm, 2x 800W/8 Ohm, 2x 2000W/2 Ohm, sym. XLR Inputs,Speaker Twist and Binding-Post Outputs, Gnd-lift, Protections, fan, 11 kg, 35,6cm depth (from front mounting rai 1 * 1 items
Catalog # IBSA139139
Supplier:  IBS Stock Items
Frequently Bought Together