Cytokeratins comprise a diverse group of intermediate filament proteins (IFPs) that are expressed in pairs in both keratinized and n on-keratinized epithelial tissue, where they constitute up to 85% of mature keratinocytes in the vertebrate epidermis 1 * 100 µG
Catalog # BWRLBS60854
Supplier: Bioworld Technology
Frequently Bought Together