The protein tyrosine phosphatase (PTP) family of proteins are signaling molecules that regulate processes such as cell growth, cell differentiation, oncogenic transformation and the mitotic cycle. 1 * 100 µG
Catalog # BWRLBS60767
Supplier: Bioworld Technology
Frequently Bought Together