Cytochrome P450 3A (CYP3A) genes encode monooxygenases—enzymes which catalyze drug metabolism and the synthesis of cholesterol, ster oids and other lipids. CYP3A, the most abundant p450 enzyme in human liver, is responsible for the metabolism of more 1 * 100 µG
Catalog # BWRLBS8464
Supplier: Bioworld Technology
Frequently Bought Together