RECOMBINANT HUMAN TUMOR NECROSIS FACTOR ALPHA (WITH HSA) 1 * 20 µG
Catalog # INVVRCYC-HTNFA
Supplier: Invivogen
RECOMBINANT HUMAN TUMOR NECROSIS FACTOR ALPHA (WITH HSA) 1 * 20 µG
Catalog # INVVRCYC-HTNFA
Supplier: Invivogen
Frequently Bought Together
Reservoir system
Supplier: Beckman Coulter
Hybridisation bottles, WHEATON®
Supplier: DWK Life Sciences
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Filter plates, 24-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Accessories and consumables for KingFisher™ Duo Prime
Supplier: Thermo Fisher Scientific
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
VWR®, Coloured Cap Inserts for Cryogenic Vials
Supplier: VWR Collection
Transfer pipettes, general purpose, Samco™
Supplier: SAMCO
VWR®, Cryo Boxes
Supplier: VWR Collection
Cap sealing mats for VersiPlate 96-well PCR strip tube plates, VersiCap™ Mats
Supplier: Thermo Fisher Scientific
Spreaders, L-shaped
Supplier: Local
Storage microplates 384-well polypropylene, Corning®
Supplier: Corning